Merck
  • Prooxidant and antioxidant action of 4-(4-phenoxybenzoyl)benzoic acid derivatives.
Prooxidant and antioxidant action of 4-(4-phenoxybenzoyl)benzoic acid derivatives.
Biopolymers (2004-03-30)
Hassan Y Aboul-Enein, Alexandra Kladna, Irena Kruk, Krzysztof Lichszteld, Mariola Marchlewicz, Teresa Michalska, Ola I A Salem, Rolf W Hartmann
ABSTRACT
4-(4-Phenoxybenzoyl)benzoic acid derivatives (PBADs) were found to inhibit rat and human alpha-reductase isozymes 1 and 2 in vitro. Chemiluminescence (CL), electron spin resonance, spin trapping techniques, and spectrophotometry were used to examine the effect of PBADs on reactive oxygen species (superoxide radical, O(2)(.-); hydroxyl radical, HO(*); singlet oxygen, (1)O(2)) generating systems. All test compounds at a concentration of 0.5 mM enhanced the CL from O(2)(.-) up to fivefold, which was recorded as the light sums during 1 min. At 0.38 mM PBAD enhanced production of HO(*) from H(2)O(2) in the presence of Co(II) up to 90%, as measured by a deoxyribose assay. Using the spin trap agent 5,5-dimethyl-1-pyrroline-N-oxide, it was found that the amplitude of the signal arising from the Fenton-like reaction [Co(II)/H(2)O(2)] was significantly diminished by the test compounds. The compounds also inhibited the (1)O(2) dependent 2,2,6,6-tetramethylpiperidine-N-oxide radical, which is generated in the acetonitrile/H(2)O(2) system. The measured rate constants of (1)O(2)-dimol quenching by PBAD were in the range of (0.8-2.6) x 10(8) M(-1) s(-1). The interaction between PBAD and (1)O(2) was also checked using a spectrophotometry method based on bleaching of p-nitrosodimethylaniline. These results indicate that PBAD may directly scavenge HO(*) and (1)O(2), but not O(2)(.-). However, the compounds that were examined had prooxidant ability under some reaction conditions.
Sigma-Aldrich
4-Phenoxybenzoic acid, 97%