Merck
  • Synthesis, crystal structure, magnetic properties, and theoretical studies of [(Cu(mepirizole)Br)2(mu-OH)(mu-pz)] (mepirizole=4-methoxy-2-(5-methoxy-3-methyl-1H-pyrazol-1-yl)-6-methylpyrimidine; pz=pyrazolate), a novel mu-pyrazolato-mu-hydroxo-dibridged copper(II) complex.
Synthesis, crystal structure, magnetic properties, and theoretical studies of [(Cu(mepirizole)Br)2(mu-OH)(mu-pz)] (mepirizole=4-methoxy-2-(5-methoxy-3-methyl-1H-pyrazol-1-yl)-6-methylpyrimidine; pz=pyrazolate), a novel mu-pyrazolato-mu-hydroxo-dibridged copper(II) complex.
Inorganic chemistry (2003-12-09)
Emilio Escrivà, Julia García-Lozano, José Martínez-Lillo, Hugo Nuñez, Juan Server-Carrió, Lucía Soto, Rosa Carrasco, Joan Cano
ABSTRACT
A novel mu-pyrazolato-mu-hydroxo-dibridged copper(II) complex has been synthesized and structurally characterized: [(Cu(mepirizole)Br)2(mu-OH)(mu-pz)] (mepirizole=4-methoxy-2-(5-methoxy-3-methyl-1H-pyrazol-1-yl)-6-methylpyrimidine; pz=pyrazolate). The title compound crystallizes in the monoclinic system, space group P2(1)/c, with a=15.618(2) A, b=15.369(3) A, c=16.071(3) A, and beta=112.250(1) degrees. The structure is built up of dinuclear [(Cu(mepirizole)Br)2(mu-OH)(mu-pz)] units with five-coordinated copper(II) ions (CuBrN3O chromophores) linked by mu2-OH and mu2-pyrazolato bridges that are well separated from each others. The intramolecular copper-copper distance is 3.378(3) A. Magnetic susceptibility data show that the copper atoms are strongly antiferromagnetically coupled with J=-770 cm(-1). The obtained triplet-singlet energy gap is compared with those reported for a series of related dimers. The strong antiferromagnetic coupling arising from the complementarity of the hydroxo and pyrazolato bridges has been discussed on the basis of DFT calculations.
Product Description
Supelco
Mepirizole, analytical standard
Pricing and availability is not currently available.