Merck
  • Human biomonitoring of pyrethrum and pyrethroid insecticides used indoors: determination of the metabolites E-cis/trans-chrysanthemumdicarboxylic acid in human urine by gas chromatography-mass spectrometry with negative chemical ionization.
Human biomonitoring of pyrethrum and pyrethroid insecticides used indoors: determination of the metabolites E-cis/trans-chrysanthemumdicarboxylic acid in human urine by gas chromatography-mass spectrometry with negative chemical ionization.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-10-03)
L Elflein, E Berger-Preiss, A Preiss, M Elend, K Levsen, G Wünsch
ABSTRACT
This work describes a gas chromatographic-mass spectrometric method employing negative chemical ionization (NCI) for the determination of E-cis/trans-chrysanthemumdicarboxylic acid (CDCA) in human urine used as a biomarker for the exposure to pyrethrum and/or certain pyrethroids in insecticide formulations applied indoors. Mixed-mode solid phase extraction was utilized for sample cleanup. Extraction recoveries ranged from 92 to 104% (2-9% R.S.D.). The acids were esterified with 1,1,1,3,3,3-hexafluoroisopropanol (HFIP) allowing both their gas chromatographic separation and their sensitive mass spectrometric detection under NCI conditions. Detection limits of ca. 0.05 microg/l urine were achieved.
Sigma-Aldrich
(+)-trans-Chrysanthemic acid, ≥97.0% (GC)