- Modulation of [3H]MK-801 binding to NMDA receptors in vivo and in vitro.
Modulation of [3H]MK-801 binding to NMDA receptors in vivo and in vitro.
[3H]MK-801 binding in vivo was used to determine the occupancy of NMDA receptor ligands shown to allosterically modulate binding in vitro. ED(50) values (mg/kg) were obtained for the channel blockers (+)-5-methyl-10,11-dihydro-5,4-dibenzo[a,d]cyclohepten-5,10-imine maleate ((+)-MK-801, 0.2), 1-(1-phenylcyclohexyl)piperidine (phencyclidine, PCP, 1.7) and ketamine (4.4). Antagonists at the glutamate (DL-(2-carboxypiperazine-4-yl)propyl-1-phosphonate (DL-CPP, 5.7)) and glycine site (7-Chloro-4-hydroxy-3-(3-phenoxy)-phenyl-2(H)quinolinone (L-701,324, 14.1), 3R(+)cis-4-methyl-pyrrollid-2-one (L-687,414, 15.1)) inhibited [3H]MK-801 binding in vivo to varying maximum levels (69%, 103% and 45%, respectively). NR2B subunit-selective compounds acting at the ifenprodil site inhibited [3H]MK-801 in vivo by a maximum of 52-72% and gave ED(50) values (mg/kg) of: (+/-)-(1S*, 2S*)-1-(4-hydroxyphenyl)-2-(4-hydroxy-4-phenylpiperidino)-1-propanol ((+/-)CP-101,606), 1.9; (+/-)-(3R, 4S)-3-[4-(4-fluorophenyl)-4-hydroxypiperidin-1-yl]chroman-4,7-diol ((+/-)CP-283,097), 1.8; (+/-)-(R*, S*)-alpha-(4-hydroxyphenyl)-beta-methyl-4-(phenylmethyl)-1-piperidine propanol ((+/-)Ro 25-6981), 1.0; ifenprodil, 6.0. The glycine site agonist D-serine stimulated binding to 151% of control with an ED(50) of 1.7 mg/kg. Results show that [3H]MK-801 binding in vivo may be used to measure receptor occupancy of ligands acting not only within the ion channel but also at modulatory sites on the NMDA receptor complex.