Merck
  • Control of Coptotermes havilandi (Isoptera: Rhinotermitidae) with hexaflumuron baits and a sensor incorporated into a monitoring and baiting program.
Control of Coptotermes havilandi (Isoptera: Rhinotermitidae) with hexaflumuron baits and a sensor incorporated into a monitoring and baiting program.
Journal of economic entomology (2000-05-29)
N Y Su, P M Ban, R H Scheffrahn
ABSTRACT
A sensor consisting of a wooden monitor painted with a conductive circuit of silver particle emulsion was placed in a monitoring station to detect feeding activity of the subterranean termite Coptotermes havilandi Holmgren. Sensor accuracy was 100% 1 mo after installation, but 9 mo after sensor placement, the rate declined to 73%. After the detection of C. havilandi in the stations, baits containing the chitin synthesis inhibitor hexaflumuron were applied in five colonies, and four colonies were eliminated within 3-5 mo. Baiting could not be completed for the remaining one colony because the site became inaccessible.
Product Description
Supelco
Hexaflumuron, PESTANAL®, analytical standard
Pricing and availability is not currently available.