Merck
  • Influence of chromium tripicolinate on growth and glucose metabolism in yearling horses.
Influence of chromium tripicolinate on growth and glucose metabolism in yearling horses.
Journal of animal science (1999-11-24)
E A Ott, J Kivipelto
ABSTRACT
Thoroughbred and Quarter Horse yearlings (n = 24; 335+/-7 d of age) were used in a 112-d feeding trial to determine whether chromium (Cr) supplementation would alter growth, development, and energy metabolism of growing horses on high-concentrate diets. The horses were assigned at random within breed and gender subgroups to one of four treatment groups: A) basal concentrate; B) basal plus 175 microg of Cr/kg concentrate; C) basal plus 350 microg of Cr/kg concentrate; and D) basal plus 700 microg of Cr/kg concentrate. Chromium was provided via Cr tripicolinate (Prince Agri Products, Quincy, IL). The horses were weighed, measured for withers and hip height, heart girth, and body length and underwent ultrasound evaluation for croup fat thickness. The concentrate was fed for ad libitum consumption for two, 1.5-hr feeding periods daily. Coastal bermudagrass (Cynodon dactylon) hay was group-fed (six animals/group) at 1% of BW daily. Feed intake was 60% concentrate and 40% hay, resulting in a supplemental Cr intake of 0, 105, 210, and 420 microg/kg diet for groups A, B, C, and D, respectively. Colts consumed more concentrate and total feed than did fillies (P < .05), but no dietary effect on feed intake was detected. Colts weighed more than fillies at the completion of the experiment (P = .0754), but no dietary effects on weight, body measurements, or croup fat were detected. An i.v. glucose tolerance test (.2 g of glucose/kg BW) and an i.v. insulin sensitivity test (.1 IU of insulin/kg BW) were conducted on each animal during the third 28-d period of the experiment. Plasma glucose peaked immediately following injection and decreased more rapidly in animals consuming the high-Cr diet than in those consuming the control diet (P < .01). Mean glucose fractional turnover rate values increased (P = .0369) and mean half-life of glucose decreased (P = .0634) in response to the high Cr supplementation. Plasma glucose depletions in animals fed the other two diets were between and not different from (P > .10) the depletions in control animals or in those fed high-Cr diets. No difference in insulin sensitivity was detected (P > .10). Results indicate that Cr tripicolinate supplementation of yearling horses increases the rate at which glucose is metabolized and may lower the plasma glucose concentration. No effect of Cr supplementation on development of the animals was detected.
Sigma-Aldrich
Chromium picolinate, ≥98% (HPLC)
Pricing and availability is not currently available.