Merck
  • High-density Escherichia coli cultures for continuous L(-)-carnitine production.
High-density Escherichia coli cultures for continuous L(-)-carnitine production.
Applied microbiology and biotechnology (1999-07-28)
J M Obón, J R Maiquez, M Cánovas, H P Kleber, J L Iborra
ABSTRACT
The use of a biological procedure for L-carnitine production as an alternative to chemical methods must be accompanied by an efficient and highly productive reaction system. Continuous L-carnitine production from crotonobetaine was studied in a cell-recycle reactor with Escherichia coli O44 K74 as biocatalyst. This bioreactor, running under the optimum medium composition (25 mM fumarate, 5 g/l peptone), was able to reach a high cell density (26 g dry weight/l) and therefore to obtain high productivity values (6.2 g L-carnitine l-1 h-1). This process showed its feasibility for industrial L-carnitine production. In addition, resting cells maintained in continuous operation, with crotonobetaine as the only medium component, kept their biocatalytic capacity for 4 days, but the biotransformation capacity decreased progressively when this particular method of cultivation was used.
Product Description
Levocarnitine impurity A, European Pharmacopoeia (EP) Reference Standard