Merck
  • Effect of chlorpyrifos-methyl on steroid and thyroid hormones in rat F0- and F1-generations.
Effect of chlorpyrifos-methyl on steroid and thyroid hormones in rat F0- and F1-generations.
Toxicology (2006-02-14)
Sang-Hee Jeong, Byung-Yong Kim, Hwan-Goo Kang, Hyun-Ok Ku, Joon-Hyoung Cho
ABSTRACT
Chlorpyrifos-methyl (CPM) suppressed androgenic activity in Hershberger assay using castrated rats. Acute oral lowest-observed-adverse-effect-level (LOAEL) and no-observed-adverse-effect-level (NOAEL) was evaluated as 12 and 0.1 mg/kg bw, respectively, based on its major effect of cholinesterase inhibition. Also, repeated oral NOAEL was 0.1 mg/kg bw/day based on adrenal damage in rats. We investigated one-generation reproductive toxicity of CPM focusing on endocrine-disrupting effects by the administration of 1, 10 and 100 mg/kg bw/day CPM to mature SD rats (F0) through pre-mating, mating, gestation and lactation period and to their offspring (F1) until 13 weeks age via gavage. A group treated with corn oil served as vehicle control. In F0 rats, the most affected organs were adrenal glands as increased in weight at all doses of CPM in males and at 10 and 100 mg/kg CPM in females and adrenal vacuolation at CPM 10 and 100 mg/kg. The relative and absolute ovaries and the absolute seminal vesicle weights were decreased but the weights of liver, spleen or kidneys were increased at 100 mg/kg CPM. Parameters representing reproductive performances as mating ratio, gestation length and delivery index were not affected, except for decreased fertility index and numbers of implantation and born pups and a higher male sex ratio of pups at CPM 100 mg/kg. F1 pups exposed to CPM 100 mg/kg in utero and via maternal milk showed lower body weight with changes of absolute or relative weights of brain, ovary, liver, spleen and epididymis and decreased absolute not relative anogenital distance at weanling time. The time of vaginal patency and preputial separation and estrous cycling pattern of F1 rats were not impacted by CPM. After further 10 weeks oral administration until 13 weeks old, adrenal glands, brain, liver, spleen or kidneys tended to be increased, while thyroid gland, testes and ventral prostate of F1 male rats were decreased at CPM 10 or 100 mg/kg. Histopathologically, necrosis or vacuolation of thyroid follicular epithelial cells and adrenal cortical cells were observed at all doses of CPM. Serum levels of estradiol, testosterone, T4 and T3 were significantly lower while TSH and cholesterol were higher in both F1 female and male rats treated with CPM though dose-responsiveness was not clear in F1 females. Decreased sperm were counted in F1 rats at CPM 100 mg/kg. As a whole, LOAEL and NOAEL was evaluated as 10 and 1 mg/kg bw, respectively, based on decreased estradiol and T4 and increased TSH in serum of F1 male rats, and when considering histopathological alteration of adrenal and thyroid glands, LOAEL assumed to be lower than 1 mg/kg bw. This study elucidates that CPM exhibit weak reproductive toxicity in F0 rats exposed at adulthood and negligible effects in F1 offspring exposed in utero and via lactation at weanling, but induce anti-androgenic effect and hypothyroidism after long term exposure from in utero through sexual maturation of F1 rats.
Product Description
Supelco
Chlorpyrifos-methyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.