Merck
  • Role of beta-oxidation enzymes in gamma-decalactone production by the yeast Yarrowia lipolytica.
Role of beta-oxidation enzymes in gamma-decalactone production by the yeast Yarrowia lipolytica.
Applied and environmental microbiology (2001-11-28)
Y Waché, M Aguedo, A Choquet, I L Gatfield, J M Nicaud, J M Belin
ABSTRACT
Some microorganisms can transform methyl ricinoleate into gamma-decalactone, a valuable aroma compound, but yields of the bioconversion are low due to (i) incomplete conversion of ricinoleate (C(18)) to the C(10) precursor of gamma-decalactone, (ii) accumulation of other lactones (3-hydroxy-gamma-decalactone and 2- and 3-decen-4-olide), and (iii) gamma-decalactone reconsumption. We evaluated acyl coenzyme A (acyl-CoA) oxidase activity (encoded by the POX1 through POX5 genes) in Yarrowia lipolytica in lactone accumulation and gamma-decalactone reconsumption in POX mutants. Mutants with no acyl-CoA oxidase activity could not reconsume gamma-decalactone, and mutants with a disruption of pox3, which encodes the short-chain acyl-CoA oxidase, reconsumed it more slowly. 3-Hydroxy-gamma-decalactone accumulation during transformation of methyl ricinoleate suggests that, in wild-type strains, beta-oxidation is controlled by 3-hydroxyacyl-CoA dehydrogenase. In mutants with low acyl-CoA oxidase activity, however, the acyl-CoA oxidase controls the beta-oxidation flux. We also identified mutant strains that produced 26 times more gamma-decalactone than the wild-type parents.
Product Description
Supelco
Methyl ricinoleate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)
Pricing and availability is not currently available.