Merck
  • Low-voltage field-emission scanning electron microscopy of non-coated guinea-pig hair cell stereocilia.
Low-voltage field-emission scanning electron microscopy of non-coated guinea-pig hair cell stereocilia.
Hearing research (1995-10-01)
E A Dunnebier, J M Segenhout, D Kalicharan, W L Jongebloed, H P Wit, F W Albers
ABSTRACT
The stereociliar structures of the guinea-pig cochlear organ of Corti were studied at low-voltage (1-5 kV) with field-emission scanning electron microscope (SEM) using various pre- and post-fixation methods, such as OTOTO (OsO4/thiocrbohydrazide/OsO4/thiocarbohydrazide/OsO4) and TAO (tannic acid/arginine/OsO4), and different dissection procedures of the cochlea. A perfusion and immersion pre-fixation with glutaraldehyde, in combination with removal of the bony wall and stria vascularis from the cochlea, followed by the TAO non-coating treatment gave the best result at 2 kV acceleration voltage. Due to these new technique, several interesting delicate structures of the stereocilia, in particular fine surface structures, were detected for the first time using SEM. These findings include the different types of cross-links and tip links, i.e., the fine surface morphology of the stereocilia and their attachments and imprints in the tectorial membrane (TM). One of the most interesting findings in this study is a network of long filamentous structures, which has been identified mainly at the top of the longest stereocilia and the undersurface of the TM and which may represent the glycocalyx. These findings and their possible implications in the process of mechanoelectrical transduction will be discussed.
Sigma-Aldrich
Thiocarbohydrazide, 98%
Pricing and availability is not currently available.