Merck
  • Inulin synthesis by a combination of purified fructosyltransferases from tubers of Helianthus tuberosus.
Inulin synthesis by a combination of purified fructosyltransferases from tubers of Helianthus tuberosus.
FEBS letters (1996-04-29)
M Lüscher, C Erdin, N Sprenger, U Hochstrasser, T Boller, A Wiemken
ABSTRACT
Sucrose-sucrose 1-fructosyltransferase (1-SST) was purified 100-fold from tubers of Helianthus tuberosus L. The purified enzyme was essentially devoid of invertase activity and could be separated by isoelectric focusing into five isoforms which all were composed of two subunits (59 and 26 kDa). Fructan-fructan 1-fructosyltransferase (1-FFT) was purified from the same source [M. Lüscher et al. (1993) New Phytologist 123, 437-442). When incubated individually with sucrose, 1-FFT was inactive while 1-SST formed isokestose (trimer) and, upon prolonged incubation, some nystose (tetramer). When a combination of the two enzymes was incubated with sucrose, a series of oligofructosides with a degree of polymerization of up to 20 was formed. Amino acid sequences of tryptic peptide fragments from both 1-SST and 1-FFT indicate that these enzymes are highly homologous with plant invertases.
Product Description
Supelco
Nystose, analytical standard