Merck
  • Study of the mechanism behind adjuvant action of diethylethoxymethylene malonate enhancing the rectal absorption of cefmetazole and lysozyme.
Study of the mechanism behind adjuvant action of diethylethoxymethylene malonate enhancing the rectal absorption of cefmetazole and lysozyme.
Journal of pharmacobio-dynamics (1984-09-01)
T Nishihata, M Miyake, A Kamada
ABSTRACT
Diethylethoxymethylene malonate (DEEMM) and diethyl maleate (DEM) enhanced the rectal absorption of cefmetazole in rats when studies were carried out using in vivo, in situ rectal loop and in vitro rectal everted sac method. Since an increase of cefmetazole transport was found when concentration of nonprotein sulfhydryls in rectal tissue was decreased by the presence of either DEEMM or DEM in the study using in vitro everted sac method and the enhancement of cefmetazole absorption by the coadministration of either DEEMM or DEM was suppressed by the treatment with dimercaprol, membrane permeability of rectal tissue seems to be regulated by the concentration of nonprotein sulfhydryls in rectal tissue. On the other hand, it was also found that there were differences between the action of DEEMM and the action of DEM from the following two results: 1) enhancing action of DEEMM was suppressed by the presence of calcium ion in microenema while action of DEM was not so much influenced, and 2) DEEMM enhanced the rectal absorption of lysozyme in rabbits but DEM did not.
Sigma-Aldrich
Diethyl ethoxymethylenemalonate, 99%
Pricing and availability is not currently available.