Merck
Short-term ammonium inhibition of nitrogen fixation in Azotobacter.
Biochemical and biophysical research communications (1984-09-17)
F J Cejudo, A de la Torre, A Paneque
ABSTRACT
Addition of NH4Cl at low concentrations to Azotobacter chroococcum cells caused an immediate cessation of nitrogenase activity, which was recovered once the added NH+4 was exhausted from the medium. In the presence of inhibitors of ammonium assimilation, such as L-methionine-DL-sulfoximine, L-methionine sulfone or 6-diazo-5-oxo-L-norleucine, externally added NH+4 had no effect on nitrogenase activity and the newly-fixed nitrogen was excreted into the medium as NH+4. It is concluded that, in A. chroococcum, NH+4 must be assimilated to exert its short-term inhibitory effect on nitrogen fixation.
Sigma-Aldrich
L-Methionine sulfone
Sigma-Aldrich
DL-Methionine sulfone, ≥99.0% (NT)