Merck
  • Molecular resolution electron micrographs of monolamellar paraffin crystals.
Molecular resolution electron micrographs of monolamellar paraffin crystals.
Science (New York, N.Y.) (1985-08-02)
F Zemlin, E Reuber, E Beckmann, E Zeitler, D L Dorset
ABSTRACT
A liquid helium-cooled cryoelectron microscope, operated to expose the specimen to only a very low electron dose, was used to obtain structural images of monolamellar n-tetratetracontane (n-C44H90) crystals at 0.25-nanometer resolution. These results are in contrast to earlier predictions that such extremely beam-sensitive materials could not be studied directly at this level of detail. Analysis of the resultant lattice images gives direct evidence for crystal bending as well as direct visualization of edge dislocations in this material.
Product Description
Supelco
Tetratetracontane, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Tetratetracontane, 99%
Pricing and availability is not currently available.