Merck
  • Ideal imaging in MR: contrast-enhancing pharmaceuticals. Evaluation of tempo carboxylic acid as nitroxide diagnostic agent.
Ideal imaging in MR: contrast-enhancing pharmaceuticals. Evaluation of tempo carboxylic acid as nitroxide diagnostic agent.
European journal of radiology (1986-02-01)
J L Lamarque, C Almès, J P Rouanet, X Prat, J M Bruel, J Pujol, M J Rodière, J M Martin, P Lopez, M Rossi
ABSTRACT
The meglumine salt of TCA, a piperidinyl nitroxide stable free radical derivative, was tested as an NMR contrast enhancer. This report of its use in animals indicates its usefulness in the enhancement of signal intensity of the kidney and bladder.
Sigma-Aldrich
4-Carboxy-TEMPO, free radical, 97%