Merck
  • Microbial degradation of xenobiotic, aromatic pollutants in humic water.
Microbial degradation of xenobiotic, aromatic pollutants in humic water.
Applied and environmental microbiology (1988-07-01)
P Larsson, L Okla, L Tranvik
ABSTRACT
The microbial degradation of a number of 14C-labeled, recalcitrant, aromatic pollutants, including trichloroguaiacol and di-, tri-, and pentachlorophenol, was investigated in aquatic model systems in the laboratory. Natural, mixed cultures of microorganisms in the water from a brown-water lake with a high content of humic compounds mineralized all of the tested substances to a higher degree than did microorganisms in the water from a clear-water lake. Dichlorophenol was the most rapidly degraded pollutant.
Sigma-Aldrich
3,4-Dichlorophenol, 99%
Pricing and availability is not currently available.