Merck
  • [Biocompatibility of microsurgical suture material modified with element-organic compounds].
[Biocompatibility of microsurgical suture material modified with element-organic compounds].
Klinicheskaia khirurgiia (1989-01-01)
S D Makeev, N A Galatenko, N N Bufius, E S Savitskaia, V N Polishchuk
ABSTRACT
By the method of tissue culture, it is established that the capron threads stained by brilliant green, coated with the chemosorbed substances on the base of titanium alkoxides, polyalkylhydridsiloxanes and polyorganilsiloxanes have no histotoxic properties and can be used in microsurgical practice.
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) butoxide, purum, ≥97.0% (gravimetric)
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) butoxide, polymer
Pricing and availability is not currently available.