Merck
  • Binding mechanism of the farnesoid X receptor marine antagonist suvanine reveals a strategy to forestall drug modulation on nuclear receptors. Design, synthesis, and biological evaluation of novel ligands.
Binding mechanism of the farnesoid X receptor marine antagonist suvanine reveals a strategy to forestall drug modulation on nuclear receptors. Design, synthesis, and biological evaluation of novel ligands.
Journal of medicinal chemistry (2013-05-10)
Francesco Saverio Di Leva, Carmen Festa, Claudio D'Amore, Simona De Marino, Barbara Renga, Maria Valeria D'Auria, Ettore Novellino, Vittorio Limongelli, Angela Zampella, Stefano Fiorucci
ABSTRACT
Here, we report suvanine, a marine sponge sesterterpene, as an antagonist of the mammalian bile acid sensor farnesoid-X-receptor (FXR). Using suvanine as a template, we shed light on the molecular bases of FXR antagonism, identifying the essential conformational changes responsible for the transition from the agonist to the antagonist form. Molecular characterization of the nuclear corepressor NCoR and coactivator Src-1 revealed that receptor conformational changes are associated with a specific dynamic of recruitment of these cofactors to the promoter of OSTα, a FXR regulated gene. Using suvanine as a novel hit, a library of semisynthetic derivatives has been designed and prepared, leading to pharmacological profiles ranging from agonism to antagonism toward FXR. Deep pharmacological evaluation demonstrated that derivative 19 represents a new chemotype of FXR modulator, whereas alcohol 6, with a simplified molecular scaffold, exhibits excellent antagonistic activity.
Sigma-Aldrich
Chenodeoxycholic acid