Merck
  • Metabolism of indium chelates attached to monoclonal antibody: minimal transchelation of indium from benzyl-EDTA chelate in vivo.
Metabolism of indium chelates attached to monoclonal antibody: minimal transchelation of indium from benzyl-EDTA chelate in vivo.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (1990-02-01)
S V Deshpande, R Subramanian, M J McCall, S J DeNardo, G L DeNardo, C F Meares
ABSTRACT
The metabolism of radiolabeled antibodies is important for radioimmunoimaging and therapy. The loss of indium-111 (111In) from the chelate can pose problems in imaging and increases the radiation dose to normal tissues. We have evaluated the loss in vivo of 111In from Lym-1-benzyl-EDTA-111In (an antibody conjugated with isothiocyanato-benzyl-EDTA) in normal mice. A monoclonal antibody (CHA 255) that binds to benzyl-EDTA-indium chelates, but not to other forms of indium, was used to measure the percent of 111In remaining in the chelate. Four days after injection, 97.4 +/- 2.2% of the 111In in the liver was still in the benzyl-EDTA chelate, as was 99.4 +/- 0.7% of the 111In in the urine, and 99.1 +/- 0.7% of the 111In in the blood. Studies in vitro indicate that a benzyl-EDTA-111In-antibody-chelate conjugate is more stable in human serum than a benzyl-DTPA-111In conjugate, and that both benzyl-chelate conjugates are much more stable than an unsubstituted DTPA conjugate.
Sigma-Aldrich
1-(4-Isothiocyanatobenzyl)ethylenediamine-N,N,N′,N′-tetraacetic acid, ~90% (HPLC)
Pricing and availability is not currently available.