Merck
  • Holoprosencephaly in clomiphene-induced pregnancy: a possible association? A case report and literature review.
Holoprosencephaly in clomiphene-induced pregnancy: a possible association? A case report and literature review.
Clinical and experimental obstetrics & gynecology (2013-03-01)
G Eleftheriou, R Butera, L Manzo
ABSTRACT
Clomiphene is widely used for inducing ovulation. Evidence for congenital abnormalities, in particular central nervous system defects (CNS-D) and in babies born from clomiphene-induced pregnancies is conflicting. The authors report a case of holoprosencephalia (HPE) in a fetus delivered from a mother receiving clomiphene.
Product Description
Supelco
Clomiphene citrate salt, analytical standard