Merck
  • Structural insight into inactivation of plasminogen activator inhibitor-1 by a small-molecule antagonist.
Structural insight into inactivation of plasminogen activator inhibitor-1 by a small-molecule antagonist.
Chemistry & biology (2013-02-27)
Zhonghui Lin, Jan K Jensen, Zebin Hong, Xiaoli Shi, Lihong Hu, Peter A Andreasen, Mingdong Huang
ABSTRACT
Plasminogen activator inhibitor-1 (PAI-1), a serpin, is the physiological inhibitor of tissue-type and urokinase-type plasminogen activators and thus also an inhibitor of fibrinolysis and tissue remodeling. It is a potential therapeutic target in many pathological conditions, including thrombosis and cancer. Several types of PAI-1 antagonist have been developed, but the structural basis for their action has remained largely unknown. Here we report X-ray crystal structure analysis of PAI-1 in complex with a small-molecule antagonist, embelin. We propose a mechanism for embelin-induced rapid conversion of PAI-1 into a substrate for its target proteases and the subsequent slow conversion of PAI-1 into an irreversibly inactivated form. Our work provides structural clues to an understanding of PAI-1 inactivation by small-molecule antagonists and an important step toward the design of drugs targeting PAI-1.
Sigma-Aldrich
Embelin, ≥98% (HPLC), powder
Pricing and availability is not currently available.