Merck
  • Acetylcholinesterase immobilized capillary reactors-tandem mass spectrometry: an on-flow tool for ligand screening.
Acetylcholinesterase immobilized capillary reactors-tandem mass spectrometry: an on-flow tool for ligand screening.
Journal of medicinal chemistry (2013-01-22)
Kenia L Vanzolini, Lucas C C Vieira, Arlene G Corrêa, Carmen L Cardoso, Quezia B Cass
ABSTRACT
The use of immobilized capillary enzyme reactors (ICERs) for online ligand screening has been adopted as a new technique for high-throughput screening (HTS). In this work, the selected target was the enzyme acetylcholinesterase (AChE), and the AChE-ICERs produced were used in a liquid chromatograph-tandem ion-trap mass spectrometer. The activity and kinetic parameters were evaluated by monitoring the choline's precursor ion (M + H)(+)m/z 104.0 and its ion fragment (C2H3OH) - (M + H)(+)m/z 60.0. The assay method was validated using the reference AChE inhibitors tacrine and galanthamine. Two new ligands, out of a library of 17 coumarin derivatives, were identified, and the half-maximal inhibitory concentration (IC50), inhibition constant (K(i)), and the inhibition mechanism were determined. A coumarin derivative with IC50 similar to tacrine was highlighted.
Sigma-Aldrich
Galanthamine hydrobromide from Lycoris sp., ≥94% (HPLC)
Pricing and availability is not currently available.