Merck
  • Creatine metabolism and safety profiles after six-week oral guanidinoacetic acid administration in healthy humans.
Creatine metabolism and safety profiles after six-week oral guanidinoacetic acid administration in healthy humans.
International journal of medical sciences (2013-01-19)
Sergej M Ostojic, Barbara Niess, Marko Stojanovic, Milos Obrenovic
ABSTRACT
Guanidinoacetic acid (GAA) is a natural precursor of creatine, yet the potential use of GAA as a nutritional additive for restoring creatine availability in humans has been limited by unclear efficacy and safety after exogenous GAA administration. The present study evaluated the effects of orally administered GAA on serum and urinary GAA, creatine and creatinine concentration, and on the occurrence of adverse events in healthy humans. Twenty-four healthy volunteers were randomized in a double-blind design to receive either GAA (2.4 grams daily) or placebo (PLA) by oral administration for 6 weeks. www.clinicaltrials.gov, identification number NCT01133899. Serum creatine and creatinine increased significantly from before to after administration in GAA-supplemented participants (P < 0.05). The proportion of participants who reported minor side effects was 58.3% in the GAA group and 45.5% in the placebo group (P = 0.68). A few participants experienced serum creatine levels above 70 µmol/L. Exogenous GAA is metabolized to creatine, resulting in a significant increase of fasting serum creatine after intervention. GAA had an acceptable side-effects profile with a low incidence of biochemical abnormalities.
Sigma-Aldrich
Guanidineacetic acid, 99%
Sigma-Aldrich
Creatine, anhydrous