Merck
  • [Effect of Pseudomonas aeruginosa exometabolites on planktonic and biofilm cultures of Escherichia coli].
[Effect of Pseudomonas aeruginosa exometabolites on planktonic and biofilm cultures of Escherichia coli].
Zhurnal mikrobiologii, epidemiologii, i immunobiologii (2013-01-10)
M V Kuznetsova, T I Karpunina, I L Maslennikova, L Iu Nesterova, V A Demakov
ABSTRACT
Study the effect of P. aeruginosa exometabolites on planktonic and biofilm cultures of bioluminescent E. coli strain. E. coli K12 TG1 (pF1 lux+ Ap(r)) recombinant bioluminescent strain, P. aeruginosa ATCC 27853 reference strain and 2 nosocomial isolates were used. Pyocyanin and pyoverdin content in supernatant of P. aeruginosa over-night cultures was evaluated according to E. Deziel et al. (2001). Planktonic and biofilm cultures of E. coli were obtained in 96-well plates (LB, statically, 37 degrees C), optical density of plankton, film biomass (OD600, OD580) and bioluminescence in plankton and biofilm were evaluated in microplate reader Infiniti M200 (Tecan, Austria). P. aeruginosa exometabolites increased the duration of lag-phase in E. coli, and short term exposition inhibited luminescence of planktonic cells. These effects are determined by bactericidal action ofpyocyanin and pyoverdin. Supernatants ofover-night cultures of P. aeruginosa inhibit formation of biofilm and disrupt the formed biofilm of E. coli. Effect of pyocyanin and pyoverdin on these processes is not established, other factors may have higher significance. Bioluminescence of E. coli K12 TGI that reflects the energetic status of the cell allows to evaluate and prognose the character of coexistence of P. aeruginosa in combined with E. coli planktonic and biofilm culture.
Sigma-Aldrich
Pyocyanin, Ready Made Solution from Pseudomonas aeruginosa, 5 mg/mL in DMSO
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)