Merck
  • Xylanase gene transcription in Trichoderma reesei is triggered by different inducers representing different hemicellulosic pentose polymers.
Xylanase gene transcription in Trichoderma reesei is triggered by different inducers representing different hemicellulosic pentose polymers.
Eukaryotic cell (2013-01-08)
Silvia Herold, Robert Bischof, Benjamin Metz, Bernhard Seiboth, Christian P Kubicek
ABSTRACT
The ascomycete Trichoderma reesei is a paradigm for the regulation and production of plant cell wall-degrading enzymes, including xylanases. Four xylanases, including XYN1 and XYN2 of glycosyl hydrolase family 11 (GH11), the GH10 XYN3, and the GH30 XYN4, were already described. By genome mining, we identified a fifth xylanase, XYN5, belonging to GH11. Transcriptional analysis reveals that the expression of all xylanases but xyn3 is induced by D-xylose, dependent on the cellulase and xylanase regulator XYR1 and negatively regulated by the carbon catabolite repressor CRE1. Impairment of D-xylose catabolism at the D-xylose reductase and xylitol dehydrogenase step strongly enhanced induction by D-xylose. Knockout of the L-xylulose reductase-encoding gene lxr3, which connects the D-xylose and L-arabinose catabolic pathways, had no effect on xylanase induction. Besides the induction by D-xylose, the T. reesei xylanases were also induced by L-arabinose, and this induction was also enhanced in knockout mutants in L-arabinose reductase (xyl1), L-arabitol dehydrogenase (lad1), and L-xylulose reductase (lxr3). Induction by L-arabinose was also XYR1 dependent. Analysis of intracellular polyols revealed accumulation of xylitol in all strains only during incubation with D-xylose and accumulation of L-arabitol only during incubation with L-arabinose. Induction by L-arabinose could be further stimulated by addition of D-xylose. We conclude that the expression of the T. reesei xylanases can be induced by both D-xylose and L-arabinose, but independently of each other and by using different inducing metabolites.
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)
Pricing and availability is not currently available.