Merck
  • [The dynamics of anxious depression under the treatment with antidepressants with different mechanisms of action].
[The dynamics of anxious depression under the treatment with antidepressants with different mechanisms of action].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2012-12-22)
N M Maksimova, O P Vertogradova
ABSTRACT
Peculiarities of the dynamics of anxious depression under the treatment with selective serotonergic antidepressants with different mechanisms of action on the serotonin reuptake were investigated. It was examined 61 patients with anxious depression (ICD-10 F32.1, F33.1, F34.1) treated with zoloft (sertraline) or coaxil (tianeptine) as a monotherapy. The following methods were used: clinical-psychopathological, psychometric (Hamilton Rating Scales for Depression and Anxiety, the Sheehan Patient-Related Anxiety Scale) and statistical analysis. The comparative investigation has shown that both zoloft and coaxil are practically equally effective in the treatment of anxious depression with some peculiarities in the dynamics of clinical parameters.
Sigma-Aldrich
Tianeptine sodium salt hydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.