Merck
  • Management of a toxic cyanobacterium bloom (Planktothrix rubescens) affecting an Italian drinking water basin: a case study.
Management of a toxic cyanobacterium bloom (Planktothrix rubescens) affecting an Italian drinking water basin: a case study.
Environmental science & technology (2012-11-22)
Sara Bogialli, Federica Nigro di Gregorio, Luca Lucentini, Emanuele Ferretti, Massimo Ottaviani, Nicola Ungaro, Pier Paolo Abis, Matteo Cannarozzi de Grazia
ABSTRACT
An extraordinary bloom of Planktothrix rubescens, which can produce microcystins (MCs), was observed in early 2009 in the Occhito basin, used even as a source of drinking water in Southern Italy. Several activities, coordinated by a task force, were implemented to assess and manage the risk associated to drinking water contaminated by cyanobacteria. Main actions were: evaluation of analytical protocols for screening and confirmatory purpose, monitoring the drinking water supply chain, training of operators, a dedicated web site for risk communication. ELISA assay was considered suitable for health authorities as screening method for MCs and to optimize frequency of sampling according to alert levels, and as internal control for the water supplier. A liquid chromatography-tandem mass spectrometric method able to quantify 9 MCs was optimized with the aim of supporting health authorities in a comprehensive risk evaluation based on the relative toxicity of different congeners. Short, medium, and long-term corrective actions were implemented to mitigate the health risk. Preoxidation with chlorine dioxide followed by flocculation and settling have been shown to be effective in removing MCs in the water treatment plant. Over two years, despite the high levels of cyanobacteria (up to 160 × 10(6) cells/L) and MCs (28.4 μg/L) initially reached in surface waters, the drinking water distribution was never limited.
Product Description
Supelco
Cylindrospermopsin solution, 10 μg/mL in H2O, PESTANAL®, analytical standard
Pricing and availability is not currently available.