Merck
  • Antiviral therapy can reverse the development of immune senescence in elderly mice with latent cytomegalovirus infection.
Antiviral therapy can reverse the development of immune senescence in elderly mice with latent cytomegalovirus infection.
Journal of virology (2012-11-02)
Mark Beswick, Annette Pachnio, Sarah N Lauder, Clive Sweet, Paul A Moss
ABSTRACT
Cytomegalovirus (CMV) infection leads to the development of adaptive and humoral immune responses that are among the largest for any pathogen, and intriguingly, the magnitude of the immune response increases with age, a phenomenon termed "memory inflation." Elevated CMV-specific immunity has been correlated with an increased mortality rate in elderly individuals and with impaired vaccination responses. The latent phase of CMV infection is characterized by intermittent episodes of subclinical viral reactivation and the production of immunogenic transcripts that may maintain memory inflation of virus-specific cytotoxic lymphocytes. However, the relative importance of CMV reactivation in the development of memory inflation is uncertain, as is the potential for antiviral treatment to reverse this effect. Here, we administered valaciclovir for up to 12 months in mice with established murine CMV (MCMV) infection. Treatment reduced the magnitude of the MCMV-specific CD8(+) T-lymphocyte response by 80%, and the residual MCMV tetramer-specific lymphocytes exhibited a less differentiated phenotype. In addition, latent MCMV infection suppressed the proportion of naïve CD8(+) T cells by 60% compared to antiviral-treated mice or MCMV-negative animals. Furthermore, treatment led to a reduction in influenza A viral loads following a challenge in elderly MCMV-infected animals and also reduced the differentiation of influenza virus-specific cytotoxic lymphocytes. These observations demonstrate that MCMV-specific memory inflation is maintained by viral replication and that therapeutic intervention could lead to improved immune function.
Sigma-Aldrich
Valacyclovir hydrochloride hydrate, ≥98% (HPLC), solid