Merck
  • Nitrofurantoin in the treatment of extended-spectrum β-lactamase-producing Escherichia coli-related lower urinary tract infection.
Nitrofurantoin in the treatment of extended-spectrum β-lactamase-producing Escherichia coli-related lower urinary tract infection.
International journal of antimicrobial agents (2012-10-16)
Meltem Isikgoz Tasbakan, Husnu Pullukcu, Oguz Resat Sipahi, Tansu Yamazhan, Sercan Ulusoy
ABSTRACT
The aim of this study was to evaluate the effect of nitrofurantoin (NFT) in the treatment of extended-spectrum β-lactamase (ESBL)-producing Escherichia coli-related lower urinary tract infection (LUTI). The hospital records of all patients aged >18 years with dysuria or problems with frequency or urgency in passing urine, >20 leukocytes/mm(3) in urine microscopy and culture-proven ESBL-producing NFT-sensitive E. coli in the urine (>10(5) CFU/mm(3)), no leukocytosis or fever and who were treated with NFT between January 2006 and May 2011 in our outpatient clinic or in the hospital were evaluated. All patients had received a NFT 50 mg capsule every 6 h for 14 days and had a control urine culture taken 7-9 days after therapy. Clinical success was defined as resolution of symptoms at the control visit, and microbiological success was defined as a sterile control urine culture. A total of 75 patients (mean±standard deviation age, 54±17 years; 45 females, 30 males, all but 14 with complicated LUTI) fulfilled the study inclusion criteria. Overall clinical and microbiological success rates were 69% (52/75) and 68% (51/75), respectively. Control urine culture performed 28-31 days after the end of therapy was available in 31/51 patients (61%) with microbiological success. Re-infection and relapse rates were 6.5% (2/31) and 3.2% (1/31), respectively. In conclusion, these results suggest that NFT may be an alternative in the treatment of ESBL-producing E. coli-related LUTI. This is the first study in which NFT was used in the treatment of LUTI due to ESBL-producing E. coli as well as in patients with complicated UTI.
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV)
Supelco
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nitrofurantoin, VETRANAL®, analytical standard