Merck
  • Neurologic outcomes and adjunctive steroids in HIV patients with severe cerebral toxoplasmosis.
Neurologic outcomes and adjunctive steroids in HIV patients with severe cerebral toxoplasmosis.
Neurology (2012-10-12)
Romain Sonneville, Matthieu Schmidt, Jonathan Messika, Ali Ait Hssain, Daniel da Silva, Isabelle F Klein, Lila Bouadma, Michel Wolff, Bruno Mourvillier
ABSTRACT
Cerebral toxoplasmosis remains a common neurologic complication in patients with AIDS. In this study, we aimed to characterize the prognosis of patients with HIV infection with severe forms of cerebral toxoplasmosis and to investigate the effects of adjunctive steroids on outcomes. We carried out a retrospective cohort study (2000-2011) on consecutive patients with cerebral toxoplasmosis admitted to the medical intensive care unit (ICU) of 5 hospitals. Functional prognosis was graded at 3 months using the modified Rankin Scale (mRS). We studied 100 patients with a CD4 cell count of 25 (8-62) cells/μL and a Glasgow Coma Scale (GCS) score of 11 (6-14). At follow-up, 51 patients had an mRS score of 0-2 (functional independence), 30 had an mRS score of 3-5 (severe disability), and 19 had an mRS score of 6 (death). Compared with other specific treatments, the use of pyrimethamine-sulfadiazine was associated with improved survival (p = 0.03). Two factors present at ICU admission were independently associated with a poor outcome (mRS score >2) at 3 months: a CD4 cell count <25 cells/μL (odds ratio [OR] 2.7, 95% confidence interval [CI] 1.1-6.7) and a GCS score ≤8 (OR 3.1, 95% CI 1.2-7.7). In patients treated with pyrimethamine-sulfadiazine, the use of adjunctive steroids to treat cerebral edema associated with focal lesions appeared safe but was not associated with better neurologic outcomes. Severe forms of cerebral toxoplasmosis in patients with HIV infection are characterized by a good prognosis in approximately 50% of cases. Profound immunodepression and impaired consciousness represent major determinants of outcome. In our study, the benefit of adjunctive steroids to treat cerebral edema could not be demonstrated.
Product Description
Supelco
Sulfadiazine, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfadiazine, 99.0-101.0%
Pricing and availability is not currently available.