Merck
  • Preparation of nano-sized graphite-supported CuO and Cu-Sn as active materials in lithium ion batteries.
Preparation of nano-sized graphite-supported CuO and Cu-Sn as active materials in lithium ion batteries.
Journal of nanoscience and nanotechnology (2012-08-02)
Dong-Won Jung, Jae-Hun Jeong, Byung-Seon Kong, Jung Kyoo Lee, Eun-Suok Oh
ABSTRACT
Nano-sized Cu-Sn and Cu oxide particles supported on ball-milled graphite were synthesized, and their electrochemical characteristics for use as anode active materials in lithium-ion batteries were investigated. The samples were also characterized via FE-SEM, XRD, and TGA. Most of the Cu oxides on BMG were monoclinic CuO crystals, whereas the Cu-Sn particles were composed of hexagonal Cu3Sn and tetragonal SnO2 crystals. These particles may contribute to an increase in the reversible capacity of lithium ion batteries.
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous
Sigma-Aldrich
Copper(I) oxide, powder, ≤7 μm, 97%