Merck
  • Biosynthesis of silver and gold nanoparticles using Bacillus licheniformis.
Biosynthesis of silver and gold nanoparticles using Bacillus licheniformis.
Methods in molecular biology (Clifton, N.J.) (2012-07-14)
Muthu Irulappan Sriram, Kalimuthu Kalishwaralal, Sangiliyandi Gurunathan
ABSTRACT
Owing to the wide-ranging applications of noble metal nanoparticles in diverse areas of science and technology, different methods have been proposed for their synthesis. Here, we describe the methods for the intracellular biosynthesis of silver and gold nanoparticles using the bacterium Bacillus licheniformis KK2 and this same procedure can be followed for other bacteria as well. The biological synthesis of nanoparticles is highly eco-friendly and possesses distinct advantages such as enhanced stability, better control over the size, shape, and monodispersity of the nanoparticles, when compared with the more traditional physical and chemical methods which often involves the use of hazardous chemicals creating environmental concern.
Sigma-Aldrich
Gold(III) chloride, 99%
Sigma-Aldrich
Gold(III) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Gold(I) chloride, 99.9% trace metals basis