Merck
Adsorption mechanism for xanthene dyes to cellulose granules.
Bioscience, biotechnology, and biochemistry (2012-06-29)
Aya Tabara, Chihiro Yamane, Masaharu Seguchi
ABSTRACT
The xanthene dyes, erythrosine, phloxine, and rose bengal, were adsorbed to charred cellulose granules. The charred cellulose granules were preliminarily steeped in ionic (NaOH, NaCl, KOH, KCl, and sodium dodecyl sulfate (SDS)), nonionic (glucose, sucrose, and ethanol), and amphipathic sucrose fatty acid ester (SFAE) solutions, and adsorption tests on the dye to the steeped and charred cellulose granules were conducted. Almost none of the dye was adsorbed when the solutions of ionic and amphipathic molecules were used, but were adsorbed in the case of steeping in the nonionic molecule solutions. Thin-layer chromatography (TLC) and the Fourier transform infra-red (FT-IR) profiles of SFAE which was adsorbed to the charred cellulose granules and extracted by ethyl ether suggested the presence of hydrophobic sites on the surface of the charred cellulose granules. We confirmed that the xanthene dyes could bind to the charred cellulose granules by ionic and hydrophobic bonds.
Sigma-Aldrich
Erythrosin extra bluish, certified by the Biological Stain Commission
Supelco
Erythrosin B, analytical standard
Sigma-Aldrich
Erythrosin B, certified by the Biological Stain Commission, Dye content ≥85 %
Sigma-Aldrich
Erythrosin extra bluish, for microscopy (Bact., Hist.)