Merck
  • Hypothalamic-pituitary-adrenal axis function in pony foals after neonatal ACTH-induced glucocorticoid overexposure.
Hypothalamic-pituitary-adrenal axis function in pony foals after neonatal ACTH-induced glucocorticoid overexposure.
Equine veterinary journal. Supplement (2012-05-19)
J K Jellyman, V L Allen, A J Forhead, N B Holdstock, A L Fowden
ABSTRACT
The effects of overexposure to glucocorticoids during early life of the foal on the subsequent HPA programming of the hypothalamic-pituitary-adrenal axis are unknown. To test the hypotheses that excess glucocorticoid exposure in early life subsequently increases both basal plasma concentrations of cortisol and the adrenocortical responsiveness to exogenous adrenocorticotropic hormone (ACTH). Foals received either saline (0.9% NaCl, n = 9) or long-acting ACTH (0.125 mg i.m. b.i.d., n = 6) for 5 days from Day 1 to increase endogenous cortisol concentrations. Long-term indwelling catheters were inserted under local anaesthesia into the jugular veins of foals aged 2 and 12 weeks. After recovery, short-acting ACTH1-24 was given as a single i.v. injection (2 microg/kg bwt) and blood samples were taken at 5-30 min intervals before and after ACTH administration to measure plasma cortisol concentrations. Basal plasma cortisol concentrations were higher in ACTH- than in saline-treated foals at age 3 weeks, but not at 13 weeks. There were no significant differences in either the time profile or the area under the cortisol curve in response to ACTH between the 2 groups. These data suggest that ACTH-induced overexposure to glucocorticoids during early post natal life of the foal does not have a programming effect on HPA axis function at 13 weeks. In foals, the effects of ACTH-induced overexposure to glucocorticoids, if any, may not become apparent until much later in life in a long-lived species such as the horse. These studies suggest that clinical and other stressful conditions that raise plasma cortisol concentrations during early life are unlikely to programme cardiovascular and metabolic function in horses in the short term.
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 1-24 human, rat, ≥97% (HPLC)
Pricing and availability is not currently available.