Merck
  • Palladium-catalyzed synthesis of 4-oxaspiro[2.4]heptanes via central attack of oxygen nucleophiles to π-allylpalladium intermediates.
Palladium-catalyzed synthesis of 4-oxaspiro[2.4]heptanes via central attack of oxygen nucleophiles to π-allylpalladium intermediates.
Organic letters (2012-04-26)
Ryo Shintani, Tomoaki Ito, Tamio Hayashi
ABSTRACT
A palladium-catalyzed decarboxylative cyclopropanation of γ-methylidene-δ-valerolactones with aromatic aldehydes has been developed to give 4-oxaspiro[2.4]heptanes with high selectivity. The site of nucleophilic attack to a π-allylpalladium intermediate has been controlled with a sterically demanding phosphine ligand. The course of the reaction is highly dependent on ligands and solvents, and selective formation of methylenetetrahydropyrans has also been realized.
Sigma-Aldrich
δ-Valerolactone, technical grade