- Application of dearomatization strategy on the synthesis of furoquinolinone and angelicin derivatives.
Application of dearomatization strategy on the synthesis of furoquinolinone and angelicin derivatives.
Organic letters (2012-04-07)
Yang Ye, Li Zhang, Renhua Fan
PMID22480214
ABSTRACT
The oxidative dearomatization of 3-(3-alkynyl-4-hydroxyphenyl)propanoic acid is combined with a cascade transition-metal catalyzed cyclization/addition/aromatization/lactamization sequence, which provides a novel approach to prepare furoquinolinone and angelicin derivatives in a convergent and efficient manner.
MATERIALS