Merck
  • Microflow-injection chemiluminescence of luminol and hypochlorite enhanced by phloxine B.
Microflow-injection chemiluminescence of luminol and hypochlorite enhanced by phloxine B.
Luminescence : the journal of biological and chemical luminescence (2012-02-23)
Xiaoli Song, Hong Shen, Xuefeng Yin, Xiuzhong Wang, Jinhua Liu
ABSTRACT
We report for the first time that the sensitivity of the luminol-hypochlorite chemiluminescence (CL) reaction was enhanced approximately 10 times by the addition of phloxine B. The maximum wavelength of CL emission shifted from 431 to 595 nm in the absence and presence, respectively, of phloxine B, suggesting that an efficient chemiluminescence resonance energy transfer occurred between a luminol donor and a phloxine B acceptor in the luminol-hypochlorite-phloxine B system. Based on this observation, a simple, rapid and sensitive microflow injection CL method, using a microchip with spiral channel configurations, was developed for the determination of hypochlorite. Under optimized conditions, a linear calibration curve (R(2) = 0.9944) over the range 0.1-10.0 µmol/L was obtained, with a detection limit of 0.025 µmol/L (S:N = 3). The relative standard deviation (RSD) was found to be 4.2% (n = 10) for 2.5 µmol/L hypochlorite. The sample consumption was only 2 μL, with a sample throughput of 90/h. The method has been used for determining trace amounts of hypochlorite in water samples with satisfactory results.
Sigma-Aldrich
Phloxine B, antibacterial fluorescent dye
Pricing and availability is not currently available.
Sigma-Aldrich
Phloxine B, Dye content ≥80 %, certified by the Biological Stain Commission
Pricing and availability is not currently available.