Merck
  • The modulatory influence of p-methoxycinnamic acid, an active rice bran phenolic acid, against 1,2-dimethylhydrazine-induced lipid peroxidation, antioxidant status and aberrant crypt foci in rat colon carcinogenesis.
The modulatory influence of p-methoxycinnamic acid, an active rice bran phenolic acid, against 1,2-dimethylhydrazine-induced lipid peroxidation, antioxidant status and aberrant crypt foci in rat colon carcinogenesis.
Chemico-biological interactions (2012-02-14)
Gunasekaran Sivagami, Venkatachalam Karthikkumar, Thangavel Balasubramanian, Namashivayam Nalini
ABSTRACT
We investigated the chemopreventive effect of p-methoxycinnamic acid (p-MCA), an active phenolic acid of rice bran, turmeric, and Kaemperfia galanga against 1,2-dimethylhydrazine-induced rat colon carcinogenesis. Male albino Wistar rats were randomly divided into six groups. Group 1 consisted of control rats that received a modified pellet diet and 0.1% carboxymethyl cellulose. The rats in Group 2 received a modified pellet diet supplemented with p-MCA [80 mg/kg body weight (b.wt.) post-orally (p.o.)] everyday. The rats in Groups 3-6 received 1,2-dimethylhydrazine (DMH) (20 mg/kg b.wt.) via subcutaneous injections once a week for the first 4 weeks; additionally, the rats in Groups 4, 5 and 6 received p-MCA at doses of 20, 40 and 80 mg/kg b.wt./day p.o., respectively, everyday for 16 weeks. The rats were sacrificed at the end of the experimental period of 16 weeks. The DMH-treated rats exhibited an increased incidence of aberrant crypt foci (ACF) development; an increased crypt multiplicity; decreased concentrations of tissue lipid peroxidation markers such as thiobarbituric acid reactive substances (TBARS), conjugated dienes (CD) and lipid hydroperoxides (LOOH); decreased levels of tissue enzymic antioxidants such as superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx), glutathione reductase (GR); and decreased levels of non-enzymic antioxidants such as reduced glutathione (GSH) and vitamins C, E and A in the colon. Supplementation with p-MCA significantly reversed these changes and significantly inhibited the formation of ACF and its multiplicity. Thus, our findings demonstrate that p-MCA exerts a strong chemopreventive activity against 1,2-dimethylhydrazine-induced colon carcinogenesis by virtue of its ability to prevent the alterations in DMH-induced circulatory and tissue oxidative stress and preneoplastic changes. p-MCA was more effective when administered at a dose of 40 mg/kg b.wt. than at the other two doses tested.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%