Merck
  • Influence of glutathione S-transferases polymorphisms on biological monitoring of exposure to low doses of benzene.
Influence of glutathione S-transferases polymorphisms on biological monitoring of exposure to low doses of benzene.
Toxicology letters (2011-12-17)
Mariella Carrieri, Giovanni Battista Bartolucci, Maria Luisa Scapellato, Giovanna Spatari, Daniela Sapienza, Leonardo Soleo, Piero Lovreglio, Giovanna Tranfo, Maurizio Manno, Andrea Trevisan
ABSTRACT
The environmental and biological monitoring of benzene exposure is crucial to prevent the toxic effects of this solvent in workers. The degree of correlation, however, between the two and of different biomarkers among them varies, particularly at low levels of exposure, depending on various factors, including variability in metabolizing enzymes and smoking habits. To investigate these further, a cohort of 28 petrochemical workers (6 smokers and 22 non smokers) was monitored throughout ten consecutive days, on two occasions, two years apart, by collecting in total 173 environmental and biological samples. The airborne benzene levels, the urinary t,t-muconic acid (t,t-MA) and S-phenylmercapturic acid (S-PMA) concentrations, and the glutathione S-transferases (GST) M1 and T1 genotypes were measured. S-PMA was the only metabolite statistically correlated with airborne benzene levels (r=0.447, P<0.0001), particularly in non smokers (r=0.667, P<0.0001), the smoking habit being the only variable influencing metabolite excretion. Finally, a reduced S-PMA excretion was found to be associated with the GSTT1, but not the GSTM1, null genotype. In conclusion, the results show that S-PMA, but not t,t-MA, is able to monitor exposure to low benzene concentrations and confirm that the GSTT1 null genotype has a significant influence on metabolite excretion. The influence of the GSTT1 null genotype, however, was low, even when studying each subject with several urine samples.
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Supelco
N-Acetyl-S-phenyl-L-cysteine, analytical standard