Merck
  • Nickel-thiolate complex catalyst assembled in one step in water for solar H2 production.
Nickel-thiolate complex catalyst assembled in one step in water for solar H2 production.
Journal of the American Chemical Society (2011-12-03)
Wei Zhang, Jindui Hong, Jianwei Zheng, Zhiyan Huang, Jianrong Steve Zhou, Rong Xu
ABSTRACT
We report the use of a simple complex assembled from Ni(II) salt and 2-mecaptoethanol in one step in water as the efficient catalyst in a molecular hydrogen system which can be sensitized by a low-cost xanthene dye, Erythrosin B. An excellent quantum efficiency of 24.5% is attained at 460 nm. This simple system is expected to contribute toward the development of economical and environmentally benign solar hydrogen production systems.
Product Description
Supelco
Erythrosin B, analytical standard
Sigma-Aldrich
Erythrosin extra bluish, for microscopy (Bact., Hist.), adsorption and fluorescent indicator
Sigma-Aldrich
Erythrosin B, certified by the Biological Stain Commission, Dye content ≥85 %
Sigma-Aldrich
Erythrosin extra bluish, certified by the Biological Stain Commission