Merck
  • Effects of CaO/P2O5 ratio on the structure and elastic properties of SiO2-CaO-Na2O-P2O5 bioglasses.
Effects of CaO/P2O5 ratio on the structure and elastic properties of SiO2-CaO-Na2O-P2O5 bioglasses.
Journal of materials science. Materials in medicine (2011-11-24)
Chung-Cherng Lin, Shih-Fan Chen, Kak Si Leung, Pouyan Shen
ABSTRACT
The evolution of elastic properties and structure upon the change of CaO/P(2)O(5) ratio in SiO(2)-CaO-Na(2)O-P(2)O(5) glasses (45S5-derived and 55S4-derived) at ambient conditions has been studied by using both Brillouin and Raman spectroscopy coupled with X-ray diffraction. Under the same SiO(2)/Na(2)O ratio, it is found that a decrease in CaO/P(2)O(5) molar ratio has caused a more-polymerized silicate network via a net consumption of Q(0), Q(1), and Q(2) species yet enriching in Q(3) and Q(4) species. Brillouin experiments revealed that all the bulk, shear and Young's moduli of the glasses studied increases with the increase of CaO/P(2)O(5) molar ratio. The unexpected variation trend in shear modulus can be correlated to the contribution from cohesion, the less-polymerized phosphate Q species, and density. Compared to the 45S5-derived, the more-polymerized 55S4-deived glass has a lower bulk but slightly higher shear modulus at the given CaO/P(2)O(5) ratio.
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Sodium oxide, 80%