Merck
  • Electropolymerized molecular imprinting on gold nanoparticle-carbon nanotube modified electrode for electrochemical detection of triazophos.
Electropolymerized molecular imprinting on gold nanoparticle-carbon nanotube modified electrode for electrochemical detection of triazophos.
Colloids and surfaces. B, Biointerfaces (2011-10-01)
Huaifen Li, Chenggen Xie, Shanqi Li, Ke Xu
ABSTRACT
An electrochemical sensor for pesticide triazophos (TAP) was prepared by deposition of gold nanoparticles (AuNPs) on carbon nanotubes (CNTs) modified glassy carbon (GC) electrode surface using a potentiostatic method, followed by electropolymerizing of o-hydroxyphenol at the AuNP/CNT/GC electrode surface in the presence of template triazophos via cyclic voltammetry. The electrochemical response of triazophos at the TAP-imprinted polyhydroxyphenol (PHP) modified AuNP/CNT/GC (PHP/AuNP/CNT/GC) electrode was investigated by cyclic voltammetry. The cyclic voltammetric response of triazophos at the TAP-imprinted PHP/AuNP/CNT/GC electrode was significantly higher than that at bare GC, CNT/GC, AuNP/CNT/GC, imprinted PHP/CNT/GC and non-imprinted PHP/AuNP/CNT/GC electrodes. The results indicated that the TAP-imprinted PHP/AuNP/CNT/GC electrode can effectively improve the reductive properties of triazophos and eliminate interferences of other pesticides. In addition, the AuNPs can strikingly amplify the electrochemical response of triazophos and improve the sensitivity to triazophos. Finally, the electrochemical sensor was successfully applied to determination of triazophos in vegetable samples with satisfactory results.