Merck
  • Fluorescent coronene monoimide gels via H-bonding induced frustrated dipolar assembly.
Fluorescent coronene monoimide gels via H-bonding induced frustrated dipolar assembly.
Chemical communications (Cambridge, England) (2011-09-29)
Ankit Jain, K Venkata Rao, Chidambar Kulkarni, Anjana George, Subi J George
ABSTRACT
Two stage self-assembly of novel coronene monoimide (CMI) based gels that results in resurfacing of monomer emission in the aggregated state is reported. This process is attributed to a frustrated head-head dipolar assembly forced by hydrogen bonding.
Sigma-Aldrich
Coronene, 97%
Sigma-Aldrich
Coronene, purified by sublimation, 99%