Merck
Stabilization of alkaline phosphatase with Au@Ag2O nanoparticles.
Langmuir : the ACS journal of surfaces and colloids (2011-08-19)
Brian A Zaccheo, Richard M Crooks
ABSTRACT
Here, we report that a conductive Au@Ag(2)O nanoparticle structure significantly enhances the stability of alkaline phosphatase (AlkP) in the presence of the inhibitors urea and l-phenylalanine (Phe). The enzyme/nanoparticle construct is prepared by associating the enzyme with citrate-capped Au particles, and then adding Ag(+). UV-vis and XPS spectroscopy and transmission electron microscopy confirm the core@shell structure. AlkP activity was quantified in the presence and absence of the two inhibitors using a time-resolved colorimetric assay. The results indicate that 21% of the initial active AlkP is incorporated into the nanoparticle structure. More importantly, however, the Au@Ag(2)O core@shell host reduces the inhibitory effect of urea and Phe by factors ranging from 3 to 12, depending on the inhibitor and its concentration, compared to the wild-type enzyme.
Sigma-Aldrich
Silver oxide, predominantly silver(II) oxide
Sigma-Aldrich
Silver(I) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Silver(I) oxide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver(I) oxide, ReagentPlus®, 99%