Merck
  • Synthesis and biological evaluation of (±)-benzhydrol derivatives as potent non-nucleoside HIV-1 reverse transcriptase inhibitors.
Synthesis and biological evaluation of (±)-benzhydrol derivatives as potent non-nucleoside HIV-1 reverse transcriptase inhibitors.
Bioorganic & medicinal chemistry (2011-07-27)
Xiao-Dong Ma, Xuan Zhang, Shi-Qiong Yang, Hui-Fang Dai, Liu-Meng Yang, Shuang-Xi Gu, Yong-Tang Zheng, Qiu-Qin He, Fen-Er Chen
ABSTRACT
A series of (±)-benzhydrol derivatives featuring the essential sulfonamide group at the para position on the C-ring were synthesized and evaluated for the potential anti-HIV activity in C8166 cells. Most of these analogues demonstrated low concentration inhibitory activity with EC(50) values less than 1 μM against the wild-type HIV-1. In particular, compound 7h was identified as the highest active inhibitor of wild-type HIV-1 with an EC(50) value of 0.12 μM and selectivity index value of 312.73. Furthermore, some of them also exhibited moderate activity against the double mutant strain A(17) (K103N+Y181C) with EC(50) values lower than 5 μM. In addition, the binding modes with RT and the preliminary structure-activity relationships of these derivatives were also explored for further chemical modifications.
Sigma-Aldrich
Diphenylmethanol, 99%