Merck
  • Monocrotophos induced apoptosis in PC12 cells: role of xenobiotic metabolizing cytochrome P450s.
Monocrotophos induced apoptosis in PC12 cells: role of xenobiotic metabolizing cytochrome P450s.
PloS one (2011-03-30)
Mahendra Pratap Kashyap, Abhishek Kumar Singh, Vivek Kumar, Vinay Kumar Tripathi, Ritesh Kumar Srivastava, Megha Agrawal, Vinay Kumar Khanna, Sanjay Yadav, Swatantra Kumar Jain, Aditya Bhushan Pant
ABSTRACT
Monocrotophos (MCP) is a widely used organophosphate (OP) pesticide. We studied apoptotic changes and their correlation with expression of selected cytochrome P450s (CYPs) in PC12 cells exposed to MCP. A significant induction in reactive oxygen species (ROS) and decrease in glutathione (GSH) levels were observed in cells exposed to MCP. Following the exposure of PC12 cells to MCP (10(-5) M), the levels of protein and mRNA expressions of caspase-3/9, Bax, Bcl(2), P(53), P(21), GSTP1-1 were significantly upregulated, whereas the levels of Bclw, Mcl1 were downregulated. A significant induction in the expression of CYP1A1/1A2, 2B1/2B2, 2E1 was also observed in PC12 cells exposed to MCP (10(-5) M), whereas induction of CYPs was insignificant in cells exposed to 10(-6) M concentration of MCP. We believe that this is the first report showing altered expressions of selected CYPs in MCP-induced apoptosis in PC12 cells. These apoptotic changes were mitochondria mediated and regulated by caspase cascade. Our data confirm the involvement of specific CYPs in MCP-induced apoptosis in PC12 cells and also identifies possible cellular and molecular mechanisms of organophosphate pesticide-induced apoptosis in neuronal cells.
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Pricing and availability is not currently available.
Sigma-Aldrich
MitoLight Mitochondrial Apoptosis Detection Kit, The MitoLight Apoptosis Detection Kit for flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker.
Pricing and availability is not currently available.