Merck
  • A pilot study on lignocelluloses to ethanol and fish feed using NMMO pretreatment and cultivation with zygomycetes in an air-lift reactor.
A pilot study on lignocelluloses to ethanol and fish feed using NMMO pretreatment and cultivation with zygomycetes in an air-lift reactor.
Bioresource technology (2011-01-21)
Patrik R Lennartsson, Claes Niklasson, Mohammad J Taherzadeh
ABSTRACT
A complete process for the production of bioethanol and fungal biomass from spruce and birch was investigated. The process included milling, pretreatment with N-methylmorpholine-N-oxide (NMMO), washing of the pretreated wood, enzymatic hydrolysis, and cultivation of the zygomycetes fungi Mucor indicus. Investigated factors included wood chip size (0.5-16 mm), pretreatment time (1-5h), and scale of the process from bench-scale to 2m high air-lift reactor. Best hydrolysis yields were achieved from wood chips below 2mm after 5h of pretreatment. Ethanol yields (mg/g wood) of 195 and 128 for spruce, and 175 and 136 for birch were achieved from bench-scale and airlift, respectively. Fungal biomass yields (mg/g wood) of 103 and 70 for spruce, and 86 and 66 for birch from bench scale and airlift respectively were simultaneously achieved. NMMO pretreatment and cultivation with M. indicus appear to be a good alternative for ethanol production from birch and spruce.
Sigma-Aldrich
4-Methylmorpholine N-oxide, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methylmorpholine N-oxide solution, 50 wt. % in H2O
Pricing and availability is not currently available.