Merck
  • Nanosegregated composites of an imidazolium salt and a layered inorganic compound: organization of both anions and cations in interlayer space.
Nanosegregated composites of an imidazolium salt and a layered inorganic compound: organization of both anions and cations in interlayer space.
Nanoscale (2010-09-14)
Yuya Oaki, Hiroyuki Ohno, Takashi Kato
ABSTRACT
The organization of an imidazolium salt in the interlayer space of a layered inorganic compound leads to the formation of nanosegregated composites.
Sigma-Aldrich
4-Imidazoleacetic acid hydrochloride, 98%