Merck
Fragrance material review on 3-methyl-2-buten-1-ol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-02-10)
D McGinty, L Jones, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 3-methyl-2-buten-1-ol when used as a fragrance ingredient is presented.
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%