Merck
Impact of vibrational excitation on the kinetics of a nascent ketene.
Physical chemistry chemical physics : PCCP (2009-12-22)
Thomas Schmierer, Wolfgang J Schreier, Florian O Koller, Tobias E Schrader, Peter Gilch
ABSTRACT
The formation and decay of a ketene intermediate photochemically formed from o-nitrobenzaldehyde has been studied by femtosecond UV/Vis and IR spectroscopy. The ketene is formed predominantly within a few 100 fs and to a minor extent within approximately 200 ps via the recombination of a triplet phased bi-radical. In tetrahydrofuran solution the ketene intermediate is seen to form a secondary intermediate with biphasic kinetics. The first phase of this decay occurs within a few picoseconds. It can be attributed to the reaction of vibrationally excited ketenes. The second phase characterized by a time constant of 2 ns is due to the reaction of thermalized molecules. In 2-butanol solution the lifetime of the thermalized ketene is only approximately 60 ps and the rapid and the slow phases of the decay start to merge.
Sigma-Aldrich
2-Nitrobenzaldehyde, 98%